1-Bromooctadecane, 96%, Thermo Scientific Chemicals
1-Bromooctadecane, 96%, Thermo Scientific Chemicals
1-Bromooctadecane, 96%, Thermo Scientific Chemicals
1-Bromooctadecane, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromooctadecane, 96%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA190021000
Price (MYR)
324.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
324.00
Add to cart
Chemical Identifiers
CAS4263-52-9
IUPAC Namesodium 2-bromoethane-1-sulfonate
Molecular FormulaC2H4BrNaO3S
InChI KeyHNFOAHXBHLWKNF-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)CCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric97.5 to 102.5 % (ex Br)
Titration Argentometric97.5 to 102.5 % (ex Br)