L(+)-Lactic acid, 90% solution in water
L(+)-Lactic acid, 90% solution in water
L(+)-Lactic acid, 90% solution in water
L(+)-Lactic acid, 90% solution in water
Thermo Scientific Chemicals
L(+)-Lactic acid, 90% solution in water
CAS: 79-33-4 | C3H6O3 | 90.078 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Glass bottle
Catalog number FSA189870010
Price (MYR)
711.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
711.00
Add to cart
Chemical Identifiers
CAS107-85-7
IUPAC Name3-methylbutan-1-amine
Molecular FormulaC5H13N
InChI KeyBMFVGAAISNGQNM-UHFFFAOYSA-N
SMILESCC(C)CCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Water=<0.5 %
Appearance (Form)Liquid
GC>=98.0 %