Erythrosin B, pure, high purity, biological stain
Erythrosin B, pure, high purity, biological stain
Erythrosin B, pure, high purity, biological stain
Erythrosin B, pure, high purity, biological stain
Thermo Scientific Chemicals
Erythrosin B, pure, high purity, biological stain
CAS: 16423-68-0 | C20H6I4Na2O5 | 879.86 g/mol
Have Questions?
Catalog NumberQuantity
FSA18946025025 g
Catalog number FSA189460250
Price (MYR)
460.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
460.00
Add to cart
Chemical Identifiers
CAS38205-60-6
IUPAC Name1-(2,4-dimethyl-1,3-thiazol-5-yl)ethan-1-one
Molecular FormulaC7H9NOS
InChI KeyBLQOKWQUTLNKON-UHFFFAOYSA-N
SMILESCC(=O)C1=C(C)N=C(C)S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Infrared spectrum1.5395-1.5445 @ 20?C
Assay (GC)≥98.5%
Indicator
RUO – Research Use Only