DL-10-Camphorsulfonic acid, sodium salt, 98%, Thermo Scientific Chemicals
DL-10-Camphorsulfonic acid, sodium salt, 98%, Thermo Scientific Chemicals
DL-10-Camphorsulfonic acid, sodium salt, 98%, Thermo Scientific Chemicals
DL-10-Camphorsulfonic acid, sodium salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-10-Camphorsulfonic acid, sodium salt, 98%, Thermo Scientific Chemicals
CAS: 34850-66-3 | C10H15NaO4S | 254.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA189120050
Price (MYR)
224.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
224.00
Add to cart
Chemical Identifiers
CAS7411-49-6
IUPAC Nametetrahydrogen [1,1'-biphenyl]-3,3',4,4'-tetramine tetrachloride
Molecular FormulaC12H18Cl4N4
InChI KeyKJDSORYAHBAGPP-UHFFFAOYSA-N
SMILES[H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NC1=CC=C(C=C1N)C1=CC=C(N)C(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay9 to 11 mg/tablet
pH6.0 to 7.5
Appearance (Color)Light grayish white
Appearance (Form)Tablet