DL-10-Camphorsulfonic acid, 98%, Thermo Scientific Chemicals
DL-10-Camphorsulfonic acid, 98%, Thermo Scientific Chemicals
DL-10-Camphorsulfonic acid, 98%, Thermo Scientific Chemicals
DL-10-Camphorsulfonic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-10-Camphorsulfonic acid, 98%, Thermo Scientific Chemicals
CAS: 5872-08-2 | C10H16O4S | 232.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA189071000
Price (MYR)
422.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
422.00
Add to cart
Chemical Identifiers
CAS615-42-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to brown
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Liquid