3-Fluoropyridine, 99%, Thermo Scientific Chemicals
3-Fluoropyridine, 99%, Thermo Scientific Chemicals
3-Fluoropyridine, 99%, Thermo Scientific Chemicals
3-Fluoropyridine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Fluoropyridine, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA188300010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS304851-95-4
IUPAC Nameyttrium(3+) tritrifluoroacetate
Molecular FormulaC6F9O6Y
InChI KeyNPATVYPXYMSPKD-UHFFFAOYSA-K
SMILES[Y+3].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.