View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 2-Dimethoxy-2-phenylacetophenone
Name Note99%
CAS24650-42-8
CAS Min %98.5
Health Hazard 1LIGHT SENSITIVE