2,2-Dimethoxy-2-phenylacetophenone, 99%, Thermo Scientific Chemicals
2,2-Dimethoxy-2-phenylacetophenone, 99%, Thermo Scientific Chemicals
2,2-Dimethoxy-2-phenylacetophenone, 99%, Thermo Scientific Chemicals
2,2-Dimethoxy-2-phenylacetophenone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2-Dimethoxy-2-phenylacetophenone, 99%, Thermo Scientific Chemicals
CAS: 24650-42-8 | C16H16O3 | 256.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
250 g
Packaging:
Glass bottle
Catalog number FSA187840500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3001-72-7
IUPAC Name2H,3H,4H,6H,7H,8H-pyrrolo[1,2-a]pyrimidine
Molecular FormulaC7H12N2
InChI KeySGUVLZREKBPKCE-UHFFFAOYSA-N
SMILESC1CN2CCCN=C2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
GC>=97.5 %
Refractive index1.5190 to 1.5210 (20°C, 589 nm)