4-O-Methyldopamine hydrochloride, 98%, Thermo Scientific Chemicals
4-O-Methyldopamine hydrochloride, 98%, Thermo Scientific Chemicals
4-O-Methyldopamine hydrochloride, 98%, Thermo Scientific Chemicals
4-O-Methyldopamine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-O-Methyldopamine hydrochloride, 98%, Thermo Scientific Chemicals
CAS: 645-33-0 | C9H13NO2·HCl | 203.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
500 mg
Packaging:
Glass bottle
Catalog number FSA187740500
Price (MYR)
409.00
Add to cart
Quantity:
50 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
409.00
Add to cart
Chemical Identifiers
CAS50606-58-1
IUPAC Name1-benzylpiperidin-3-one hydrochloride
Molecular FormulaC12H16ClNO
InChI KeyOVWSFXNSJDMRPV-UHFFFAOYSA-N
SMILESCl.O=C1CCCN(CC2=CC=CC=C2)C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Cream to pale brown
Assay (Titration ex Chloride)≥94.0 to ≤106.0%
Water Content (Karl Fischer Titration)≤8.0%