1-Bromotetradecane, 98%, Thermo Scientific Chemicals
1-Bromotetradecane, 98%, Thermo Scientific Chemicals
1-Bromotetradecane, 98%, Thermo Scientific Chemicals
1-Bromotetradecane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromotetradecane, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA187381000
Price (MYR)
453.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
453.00
Add to cart
Chemical Identifiers
CAS32815-70-6
IUPAC Name2-methylhex-5-en-3-ol
Molecular FormulaC7H14O
InChI KeyJKGMJZOZIJHPOH-UHFFFAOYNA-N
SMILESCC(C)C(O)CC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4315-1.4365 @ 20°C
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥96.0%