O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 98%, C7H4F5NO.HCl, CAS Number-57981-02-9
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA186710050
Price (MYR)
4,227.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
4,227.00
Add to cart
Chemical Identifiers
CAS96-41-3
IUPAC Namecyclopentanol
Molecular FormulaC5H10O
InChI KeyXCIXKGXIYUWCLL-UHFFFAOYSA-N
SMILESOC1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4520 to 1.4540 (20°C, 589 nm)