Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
CAS: 144-62-7 | C2H2O4 | 90.034 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
View all
Plastic bottle
Catalog number FSA186430010
Price (MYR)
714.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
714.00
EA
Add to cart
Chemical Identifiers
CAS1120-71-4
IUPAC Name1,2λ⁶-oxathiolane-2,2-dione
Molecular FormulaC3H6O3S
InChI KeyFSSPGSAQUIYDCN-UHFFFAOYSA-N
SMILESO=S1(=O)CCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting solid or liquid
Infrared spectrumConforms
Appearance (Color)White to light yellow
Melting point28°C to 33°C
GC>=96.0 %
Suitable for battery materials development.
RUO – Research Use Only