1,3-Dimethyl-2-imidazolidinone, 98%, Thermo Scientific Chemicals
1,3-Dimethyl-2-imidazolidinone, 98%, Thermo Scientific Chemicals
1,3-Dimethyl-2-imidazolidinone, 98%, Thermo Scientific Chemicals
1,3-Dimethyl-2-imidazolidinone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3-Dimethyl-2-imidazolidinone, 98%, Thermo Scientific Chemicals
CAS: 80-73-9 | C5H10N2O | 114.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
Packaging:
Glass bottle
Catalog number FSA185930010
Price (MYR)
1,729.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,729.00
Add to cart
Chemical Identifiers
CAS529-19-1
IUPAC Name2-methylbenzonitrile
Molecular FormulaC8H7N
InChI KeyNWPNXBQSRGKSJB-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.5269 to 1.5289 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=97.5 %