Plumbagin, 99+%
Plumbagin, 99+%
Plumbagin, 99+%
Thermo Scientific Chemicals
Plumbagin, 99+%
CAS: 481-42-5 | C11H8O3 | 188.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA1840400101 gGlass bottle
Catalog number FSA184040010
Price (MYR)
1,079.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,079.00
Add to cart
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Appearance (Form)Granular solid
Iodine number>=1000 mg/g
Molasses number>=230
Abrasion number>=75