1H-Benzotriazole, 99%, Thermo Scientific Chemicals
1H-Benzotriazole, 99%, Thermo Scientific Chemicals
1H-Benzotriazole, 99%, Thermo Scientific Chemicals
1H-Benzotriazole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1H-Benzotriazole, 99%, Thermo Scientific Chemicals
CAS: 95-14-7 | C6H5N3 | 119.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number FSA183880025
Price (MYR)
2,780.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
2,780.00
Add to cart
Chemical Identifiers
CAS3164-29-2
IUPAC Name(2R,3R)-2,3-dihydroxybutanedioic acid diamine
Molecular FormulaC4H12N2O6
InChI KeyNGPGDYLVALNKEG-OLXYHTOASA-N
SMILESN.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Assay (Titration)> 97.5%
Chloride content5 ppm max
CommentPhosphate content: 5 ppm max
CommentSulfate: 0.005% max
View more
Indicator
RUO – Research Use Only