N,N-Dimethylformamide-d7, for NMR, 99.5% atom D, Thermo Scientific Chemicals
N,N-Dimethylformamide-d7, for NMR, 99.5% atom D, Thermo Scientific Chemicals
N,N-Dimethylformamide-d7, for NMR, 99.5% atom D, Thermo Scientific Chemicals
N,N-Dimethylformamide-d7, for NMR, 99.5% atom D, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylformamide-d7, for NMR, 99.5% atom D, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
1 mL
5 mL
Packaging:
Glass Bottle
Catalog number FSA183600100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS402-54-0
IUPAC Name1-nitro-4-(trifluoromethyl)benzene
Molecular FormulaC7H4F3NO2
InChI KeyXKYLCLMYQDFGKO-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
FormCrystals or powder or crystalline powder or lumps or fused solid
Assay (GC)≥97.5%
Melting Point (clear melt)37.0-44.0?C
For NMR
RUO – Research Use Only