Poly(vinyl alcohol), 98.0%-98.8% hydrolyzed, M.W. approx. 50,000-85,000, Thermo Scientific Chemicals
Poly(vinyl alcohol), 98.0%-98.8% hydrolyzed, M.W. approx. 50,000-85,000, Thermo Scientific Chemicals
Poly(vinyl alcohol), 98.0%-98.8% hydrolyzed, M.W. approx. 50,000-85,000, Thermo Scientific Chemicals
Poly(vinyl alcohol), 98.0%-98.8% hydrolyzed, M.W. approx. 50,000-85,000, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Poly(vinyl alcohol), 98.0%-98.8% hydrolyzed, M.W. approx. 50,000-85,000, Thermo Scientific Chemicals
Indicator
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
Catalog number FSA183380010
Price (MYR)
986.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
986.00
Add to cart
Chemical Identifiers
CAS13325-10-5
IUPAC Name4-hydroxybutan-1-aminium
Molecular FormulaC4H12NO
InChI KeyBLFRQYKZFKYQLO-UHFFFAOYSA-O
SMILES[NH3+]CCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Refractive index1.4580 to 1.4630 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow