Poly(vinyl acetate), approx. M.W. 100,000
Poly(vinyl acetate), approx. M.W. 100,000
Poly(vinyl acetate), approx. M.W. 100,000
Poly(vinyl acetate), approx. M.W. 100,000
Poly(vinyl acetate), approx. M.W. 100,000
Thermo Scientific Chemicals
Poly(vinyl acetate), approx. M.W. 100,000
CAS: 9003-20-7 | (C4H6O2)n | 86.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA183265000500 gGlass bottle
Catalog number FSA183265000
Price (MYR)
944.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
944.00
Add to cart
Chemical Identifiers
CAS7440-43-9
IUPAC Namecadmium(2+) ion
Molecular FormulaCd
InChI KeyWLZRMCYVCSSEQC-UHFFFAOYSA-N
SMILES[Cd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.002% max.
Suitable for battery materials development.
RUO – Research Use Only