15-Crown-5, 98%, Thermo Scientific Chemicals
15-Crown-5, 98%, Thermo Scientific Chemicals
15-Crown-5, 98%, Thermo Scientific Chemicals
15-Crown-5, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
15-Crown-5, 98%, Thermo Scientific Chemicals
CAS: 33100-27-5 | C10H20O5 | 220.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA182860250
Price (MYR)
733.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
733.00
Add to cart
Chemical Identifiers
CAS10031-53-5
IUPAC Nameeuropium(3+) hexahydrate trinitrate
Molecular FormulaEuH12N3O15
InChI KeyJVYYYCWKSSSCEI-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Eu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Chelation/Complexation reagent
RUO – Research Use Only