Cyclobutanemethanol, 99%, Thermo Scientific Chemicals
Cyclobutanemethanol, 99%, Thermo Scientific Chemicals
Cyclobutanemethanol, 99%, Thermo Scientific Chemicals
Cyclobutanemethanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclobutanemethanol, 99%, Thermo Scientific Chemicals
CAS: 4415-82-1 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA182230250
Price (MYR)
1,186.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,186.00
Add to cart
Specifications
Chemical Name or MaterialCyclobutanemethanol
Name Note99%
CAS4415-82-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Flammable liquid and vapor.