3-Bromobenzyl bromide, 95%, Thermo Scientific Chemicals
3-Bromobenzyl bromide, 95%, Thermo Scientific Chemicals
3-Bromobenzyl bromide, 95%, Thermo Scientific Chemicals
3-Bromobenzyl bromide, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Bromobenzyl bromide, 95%, Thermo Scientific Chemicals
CAS: 823-78-9 | C7H6Br2 | 249.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA181780050
Price (MYR)
264.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
264.00
Add to cart
Chemical Identifiers
CAS79-30-1
IUPAC Name2-methylpropanoyl chloride
Molecular FormulaC4H7ClO
InChI KeyDGMOBVGABMBZSB-UHFFFAOYSA-N
SMILESCC(C)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4055-1.4095 @ 20?C
Assay (GC)≥97.5%