Acetone-d6, for NMR, contains 1 v/v% TMS, 99.5% atom D, Thermo Scientific Chemicals
Acetone-d6, for NMR, contains 1 v/v% TMS, 99.5% atom D, Thermo Scientific Chemicals
Acetone-d6, for NMR, contains 1 v/v% TMS, 99.5% atom D, Thermo Scientific Chemicals
Acetone-d6, for NMR, contains 1 v/v% TMS, 99.5% atom D, Thermo Scientific Chemicals
Acetone-d6, for NMR, contains 1 v/v% TMS, 99.5% atom D, Thermo Scientific Chemicals
CAS: 666-52-4 | C3H6O | 64.117 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
Packaging:
Glass Bottle
Catalog number FSA181160100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS3842-55-5
IUPAC Name2-chloro-4,6-diphenyl-1,3,5-triazine
Molecular FormulaC15H10ClN3
InChI KeyDDGPPAMADXTGTN-UHFFFAOYSA-N
SMILESClC1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder
Assay (GC)≥96.0%
Melting Point (clear melt)134.0-143.0?C
For NMR
RUO – Research Use Only