N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
5 L
Packaging:
View all
Glass Bottle
Catalog number FSA181070025
Price (MYR)
806.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
806.00
Add to cart
Chemical Identifiers
CAS5390-04-5
IUPAC Namepent-4-yn-1-ol
Molecular FormulaC5H8O
InChI KeyCRWVOXFUXPYTRK-UHFFFAOYSA-N
SMILESOCCCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥94.0%
Water Content (Karl Fischer Titration)≤1.5%
Refractive Index1.4425-1.4475 @ 20°C