N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylacetamide, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
5 L
Packaging:
View all
Glass Bottle
Catalog number FSA181070010
Price (MYR)
423.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
423.00
Add to cart
Chemical Identifiers
CAS38422-62-7
IUPAC Name5-tert-butyl-2-methylfuran-3-carboxylate
Molecular FormulaC10H13O3
InChI KeyNWWAPYOVUAEIJB-UHFFFAOYSA-M
SMILESCC1=C(C=C(O1)C(C)(C)C)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Cream
Melting Point (clear melt)90.5-96.5?C
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%