Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA181051000
Price (MYR)
272.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
272.00
Add to cart
Chemical Identifiers
CAS5271-38-5
IUPAC Name2-(methylsulfanyl)ethan-1-ol
Molecular FormulaC3H8OS
InChI KeyWBBPRCNXBQTYLF-UHFFFAOYSA-N
SMILESCSCCO
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4930-1.4960 @ 20?C
Assay (GC)>98.5%
Appearance (Color)Clear colorless to pale yellow