Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dicyclohexylamine, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
Packaging:
Glass bottle
Catalog number FSA181050025
Price (MYR)
556.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
556.00
Add to cart
Chemical Identifiers
CAS136434-77-0
IUPAC Name1-bromo-2-fluoro-4-iodobenzene
Molecular FormulaC6H3BrFI
InChI KeyOCODJNASCDFXSR-UHFFFAOYSA-N
SMILESFC1=C(Br)C=CC(I)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)34.0-43.0?C
Assay (GC)≥96.0%
Appearance (Color)White to brown to pale gray
FormCrystals or powder or crystalline powder or fused solid