Cyanuric acid, 98%, Thermo Scientific Chemicals
Cyanuric acid, 98%, Thermo Scientific Chemicals
Cyanuric acid, 98%, Thermo Scientific Chemicals
Cyanuric acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyanuric acid, 98%, Thermo Scientific Chemicals
CAS: 108-80-5 | C3H3N3O3 | 129.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA181020250
Price (MYR)
208.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
208.00
Add to cart
Chemical Identifiers
CAS1308-38-9
IUPAC Namedichromium(3+) trioxidandiide
Molecular FormulaCr2O3
InChI KeyUOUJSJZBMCDAEU-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Cr+3].[Cr+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.4% max.