Cyanuric acid, 98%, Thermo Scientific Chemicals
Cyanuric acid, 98%, Thermo Scientific Chemicals
Cyanuric acid, 98%, Thermo Scientific Chemicals
Cyanuric acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyanuric acid, 98%, Thermo Scientific Chemicals
CAS: 108-80-5 | C3H3N3O3 | 129.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
250 g
Packaging:
Plastic bottle
Catalog number FSA181020010
Price (MYR)
1,378.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,378.00
Add to cart
Chemical Identifiers
CAS109-63-7
IUPAC Nameethoxyethane; trifluoroborane
Molecular FormulaC4H10BF3O
InChI KeyKZMGYPLQYOPHEL-UHFFFAOYSA-N
SMILESFB(F)F.CCOCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to straw yellow
Refractive index1.3429 to 1.3449 (20°C, 589 nm)
Assay47.5 to 49 % (BF3)