4-chlorophenol, 99+%, Thermo Scientific Chemicals
4-chlorophenol, 99+%, Thermo Scientific Chemicals
4-chlorophenol, 99+%, Thermo Scientific Chemicals
4-chlorophenol, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-chlorophenol, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA181005000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS498-60-2
IUPAC Namefuran-3-carbaldehyde
Molecular FormulaC5H4O2
InChI KeyAZVSIHIBYRHSLB-UHFFFAOYSA-N
SMILESO=CC1=COC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear yellow to brown
Infrared spectrumConforms
Refractive index1.4930 to 1.4950 (20°C, 589 nm)
GC>=97.5 %