Naphthalene, 99%, Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
CAS: 91-20-3 | C10H8 | 128.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
2.5 kg
Packaging:
View all
Plastic drum
Catalog number FSA180900050
Price (MYR)
1,032.00
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
1,032.00
Add to cart
Chemical Identifiers
CAS380430-49-9
IUPAC Name(4-{[(tert-butoxy)carbonyl]amino}phenyl)boronic acid
Molecular FormulaC11H16BNO4
InChI KeyUBVOLHQIEQVXGM-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NC1=CC=C(C=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
FormPowder
Proton NMRConforms to structure
Appearance (Color)White to cream