Naphthalene, 99%, Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
CAS: 91-20-3 | C10H8 | 128.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA180900025
Price (MYR)
645.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
645.00
Add to cart
Specifications
Chemical Name or MaterialNaphthalene
CAS91-20-3
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0