Naphthalene, 99%, Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Naphthalene, 99%, Thermo Scientific Chemicals
CAS: 91-20-3 | C10H8 | 128.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
2.5 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA180900010
Price (MYR)
404.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
404.00
Add to cart
Chemical Identifiers
CAS75-15-0
IUPAC Namemethanedithione
Molecular FormulaCS2
InChI KeyQGJOPFRUJISHPQ-UHFFFAOYSA-N
SMILESS=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Residue after evaporation=<0.002 %
Hydrogen sulfide (H2S)=<1.5 ppm
Water=<0.05 %
Appearance (Form)Liquid