Pyrene, 98%, Thermo Scientific Chemicals
Pyrene, 98%, Thermo Scientific Chemicals
Pyrene, 98%, Thermo Scientific Chemicals
Pyrene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyrene, 98%, Thermo Scientific Chemicals
CAS: 129-00-0 | C16H10 | 202.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number FSA180831000
Price (MYR)
611.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
611.00
Add to cart
Chemical Identifiers
CAS7250-53-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown to pale gray
FormPowder
Assay (Non-aqueous acid-base Titration)≥97.0 to ≤103.0%
Assay (HPLC)≥97.0%
Melting Point (clear melt)295.0-305.0?C