Pyrene, 98%, Thermo Scientific Chemicals
Pyrene, 98%, Thermo Scientific Chemicals
Pyrene, 98%, Thermo Scientific Chemicals
Pyrene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyrene, 98%, Thermo Scientific Chemicals
CAS: 129-00-0 | C16H10 | 202.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA180830250
Price (MYR)
377.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
377.00
Add to cart
Specifications
Chemical Name or MaterialPyrene
CAS129-00-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Very toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Avoid release to the environment.