Terephthalic acid, 99+%, Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
CAS: 100-21-0 | C8H6O4 | 166.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA180722500
Price (MYR)
234.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
234.00
Add to cart
Chemical Identifiers
CAS7439-89-6
IUPAC Nameiron
Molecular FormulaFe
InChI KeyXEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES[Fe]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey
Nickel (Ni)=<500 ppm
Sulfur (S)=<200 ppm
Appearance (Form)Powder
Copper (Cu)=<100 ppm