Terephthalic acid, 99+%, Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
CAS: 100-21-0 | C8H6O4 | 166.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
500 g
Packaging:
Plastic bottle
Catalog number FSA180720010
Price (MYR)
469.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
469.00
Add to cart
Chemical Identifiers
CAS223463-14-7
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Proton NMRConforms to structure
Assay (HPLC)≥94.0%
Appearance (Color)Cream
Assay (Aqueous acid-base Titration)≥94.0%