Sodium cyanate, 85%, pure
Sodium cyanate, 85%, pure
Sodium cyanate, 85%, pure
Sodium cyanate, 85%, pure
Thermo Scientific Chemicals
Sodium cyanate, 85%, pure
CAS: 917-61-3 | CNNaO | 65.01 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA180562500250 gPlastic bottle
Catalog number FSA180562500
Price (MYR)
345.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
345.00
Add to cart
Chemical Identifiers
CAS7647-17-8
IUPAC Namecaesium(1+) chloride
Molecular FormulaClCs
InChI KeyAIYUHDOJVYHVIT-UHFFFAOYSA-M
SMILES[Cl-].[Cs+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Appearance (Color)White
Titration Argentometric99.0 to 101.0 %