Phenylarsine oxide, 97%, Thermo Scientific Chemicals
Phenylarsine oxide, 97%, Thermo Scientific Chemicals
Phenylarsine oxide, 97%, Thermo Scientific Chemicals
Phenylarsine oxide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenylarsine oxide, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA180350050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPhenylarsine oxide
CAS637-03-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Health Hazard 3GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.