tert-Butyl hydroperoxide, 70% Solution in water, Thermo Scientific Chemicals
tert-Butyl hydroperoxide, 70% Solution in water, Thermo Scientific Chemicals
tert-Butyl hydroperoxide, 70% Solution in water, Thermo Scientific Chemicals
tert-Butyl hydroperoxide, 70% Solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butyl hydroperoxide, 70% Solution in water, Thermo Scientific Chemicals
CAS: 75-91-2 | C4H10O2 | 90.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA180342500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7775-27-1
IUPAC Namedisodium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular FormulaNa2O8S2
InChI KeyCHQMHPLRPQMAMX-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S(=O)(=O)OOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder or crystals
Titration Iodometric98.0 to 102.0 %
Water=<0.15 %
Heavy metals (as Pb)=<20 ppm