Poly(vinyl alcohol), 87.0-89.0% hydrolyzed, M.W. approx. 13,000-23,000
Poly(vinyl alcohol), 87.0-89.0% hydrolyzed, M.W. approx. 13,000-23,000
Poly(vinyl alcohol), 87.0-89.0% hydrolyzed, M.W. approx. 13,000-23,000
Poly(vinyl alcohol), 87.0-89.0% hydrolyzed, M.W. approx. 13,000-23,000
Thermo Scientific Chemicals
Poly(vinyl alcohol), 87.0-89.0% hydrolyzed, M.W. approx. 13,000-23,000
CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol
Have Questions?
Catalog NumberQuantity
FSA180301000100 g
Catalog number FSA180301000
Price (MYR)
276.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
276.00
Add to cart
Chemical Identifiers
CAS7704-34-9
IUPAC Namesulfur
Molecular FormulaS
InChI KeyNINIDFKCEFEMDL-UHFFFAOYSA-N
SMILES[S]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.5% minimum
Particle Size-100 mesh (less than 150 micron)