Petroleum ether, ACS reagent, boiling range 40-60°C, Thermo Scientific Chemicals
Petroleum ether, ACS reagent, boiling range 40-60°C, Thermo Scientific Chemicals
Petroleum ether, ACS reagent, boiling range 40-60°C, Thermo Scientific Chemicals
Petroleum ether, ACS reagent, boiling range 40-60°C, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Petroleum ether, ACS reagent, boiling range 40-60°C, Thermo Scientific Chemicals
CAS: 64742-49-0 | C6H14 | 86.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA180210010
Price (MYR)
529.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
529.00
Add to cart
Chemical Identifiers
CAS288-88-0
IUPAC Name1H-1,2,4-triazole
Molecular FormulaC2H3N3
InChI KeyNSPMIYGKQJPBQR-UHFFFAOYSA-N
SMILESN1C=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder and/or flakes
Infrared spectrumConforms
Melting point119°C to 122°C
Appearance (Color)White
HPLC>=99.4 %