Naphthalene, 99+%, scintillation grade, Thermo Scientific Chemicals
Naphthalene, 99+%, scintillation grade, Thermo Scientific Chemicals
Naphthalene, 99+%, scintillation grade, Thermo Scientific Chemicals
Naphthalene, 99+%, scintillation grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Naphthalene, 99+%, scintillation grade, Thermo Scientific Chemicals
CAS: 91-20-3 | C10H8 | 128.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
1 kg
Packaging:
Glass Bottle
Catalog number FSA180202500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS25013-16-5
IUPAC Name2-tert-butyl-4-methoxyphenol
Molecular FormulaC11H16O2
InChI KeyMRBKEAMVRSLQPH-UHFFFAOYSA-N
SMILESCOC1=CC=C(O)C(=C1)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
FormCrystals or powder or crystalline powder or flakes
Assay (GC)≥95.0%
For fluorescence/luminescence spectroscopy
RUO – Research Use Only