Naphthalene, 99+%, scintillation grade, Thermo Scientific Chemicals
Naphthalene, 99+%, scintillation grade, Thermo Scientific Chemicals
Naphthalene, 99+%, scintillation grade, Thermo Scientific Chemicals
Naphthalene, 99+%, scintillation grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Naphthalene, 99+%, scintillation grade, Thermo Scientific Chemicals
CAS: 91-20-3 | C10H8 | 128.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
Packaging:
Glass Bottle
Catalog number FSA180200050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS4430-20-0
IUPAC Name3,3-bis(3-chloro-4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular FormulaC19H12Cl2O5S
InChI KeyWWAABJGNHFGXSJ-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1Cl)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
lambda-max (UV-Vis)@ pH4.8: 435-442nm
Appearance (Color)Brown to dark brown to dark red
FormCrystals or powder or crystalline powder
CommentColour transition pH 4.8 (yellow / brown) to pH 6.4 (red/violet)
Loss on Drying≤15%
View more
For fluorescence/luminescence spectroscopy
RUO – Research Use Only