2'-Fluoroacetophenone, 97%
2'-Fluoroacetophenone, 97%
2'-Fluoroacetophenone, 97%
2'-Fluoroacetophenone, 97%
2'-Fluoroacetophenone, 97%
CAS: 445-27-2 | C8H7FO | 138.14 g/mol
Have Questions?
Catalog number FSA179780250
Price (MYR)
1,040.00
Add to cart
Request bulk or custom format
Price (MYR)
1,040.00
Add to cart
Chemical Identifiers
CAS129-64-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Acid-base back titration≥95.0 to <105.0%
Melting Point163-168°C