2'-Chloroacetophenone, 97%, Thermo Scientific Chemicals
2'-Chloroacetophenone, 97%, Thermo Scientific Chemicals
2'-Chloroacetophenone, 97%, Thermo Scientific Chemicals
2'-Chloroacetophenone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2'-Chloroacetophenone, 97%, Thermo Scientific Chemicals
CAS: 2142-68-9 | C8H7ClO | 154.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA179770250
Price (MYR)
462.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
462.00
Add to cart
Chemical Identifiers
CAS13478-00-7
IUPAC Namenickel(2+) hexahydrate dinitrate
Molecular FormulaH12N2NiO12
InChI KeyAOPCKOPZYFFEDA-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric>=98.5 %
Cadmium (Cd)=<5 ppm
Iron (Fe)=<10 ppm
Chromium (Cr)=<5 ppm
Aluminium (Al)=<10 ppm