Check Order Status
Methyl 1-benzyl-5-oxo-3-pyrrolidine-carboxylate, 98%, Thermo Scientific™
Element Search
Structure Search
Methyl 1-benzyl-5-oxo-3-pyrrolidine-carboxylate, 98%, Thermo Scientific™
Have Questions?
Catalog Number
Quantity
Packaging
FSA179740050
5 g
Glass bottle
Catalog number
FSA179740050
Price (MYR)
100.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
100.00
Add to cart
Specifications
Chemical Name or Material
Methyl 1-benzyl-5-oxo-3-pyrrolidine-carboxylate
Name Note
98%
CAS
51535-00-3
Melting Point
62°C to 66°C
Color
White to Yellow