2-Bromonaphthalene, 99%, Thermo Scientific Chemicals
2-Bromonaphthalene, 99%, Thermo Scientific Chemicals
2-Bromonaphthalene, 99%, Thermo Scientific Chemicals
2-Bromonaphthalene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromonaphthalene, 99%, Thermo Scientific Chemicals
CAS: 580-13-2 | C10H7Br | 207.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA179710250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-66-6
IUPAC Namezinc
Molecular FormulaZn
InChI KeyHCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey
Appearance (Form)Powder
Total trace metal impurities=<0.001 %