Naphazoline hydrochloride, 99%, Thermo Scientific Chemicals
Naphazoline hydrochloride, 99%, Thermo Scientific Chemicals
Naphazoline hydrochloride, 99%, Thermo Scientific Chemicals
Naphazoline hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Naphazoline hydrochloride, 99%, Thermo Scientific Chemicals
CAS: 550-99-2 | C14H14N2·ClH | 246.74 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number FSA179660250
Price (MYR)
534.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
534.00
Add to cart
Specifications
Chemical Name or MaterialNaphazoline hydrochloride
Name Note99%
CAS550-99-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger