Phenylselenyl chloride, 98%, Thermo Scientific Chemicals
Phenylselenyl chloride, 98%, Thermo Scientific Chemicals
Phenylselenyl chloride, 98%, Thermo Scientific Chemicals
Phenylselenyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenylselenyl chloride, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
Packaging:
Glass bottle
Catalog number FSA179510250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS622-95-7
IUPAC Name1-(bromomethyl)-4-chlorobenzene
Molecular FormulaC7H6BrCl
InChI KeyKQNBRMUBPRGXSL-UHFFFAOYSA-N
SMILESClC1=CC=C(CBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals and/or chunks or fused solid
GC>=97.5 %
Melting point46°C to 51°C
Appearance (Color)White to almost white
Infrared spectrumConforms
C-X bond formation reagent
RUO – Research Use Only