2-(Aminomethyl)-1-ethylpyrrolidine, 95%, Thermo Scientific Chemicals
2-(Aminomethyl)-1-ethylpyrrolidine, 95%, Thermo Scientific Chemicals
2-(Aminomethyl)-1-ethylpyrrolidine, 95%, Thermo Scientific Chemicals
2-(Aminomethyl)-1-ethylpyrrolidine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-(Aminomethyl)-1-ethylpyrrolidine, 95%, Thermo Scientific Chemicals
CAS: 26116-12-1 | C7H16N2 | 128.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA179480100
Price (MYR)
908.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
908.00
Add to cart
Specifications
Chemical Name or Material2-(Aminomethyl)-1-ethylpyrrolidine
Name Note95%
CAS26116-12-1
CAS Min %94.0
Health Hazard 1GHS Signal Word: Danger